Learn More
NANOMICRO TECHNOLOGIES INC NanoChrom Technologies, BioCore SC x LC Analytical Column, 5um, 4.6 x 100 mm, PEEK
Catalog No. NC3585554
Encompass_Preferred
BioCore SCX is a family of high-performance, strong cation-exchange columns designed for separating charged variants in proteins, including monoclonal antibodies (mAbs). Its column technology involves the creation of a hydrophilic sulfonic functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.
Catalog No. NC3585554 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. B41105000004610P
BioCore SCX is a family of high-performance, strong cation-exchange columns designed for separating charged variants in proteins, including monoclonal antibodies (mAbs). Its column technology involves the creation of a hydrophilic sulfonic functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.