Learn More
NANOMICRO TECHNOLOGIES INC NanoChrom Technologies, BioCore SEC-150 LC Analytical Column, 3um, 4.6 x 150 mm, Stainless Steel
Catalog No. NC3585536
Encompass_Preferred
BioCore SEC is a family of high performance, size exclusion chromatography columns designed for separating a broad range of biomolecules based on size. This column technology involves the creation of a neutral hydrophilic layer on the surface of specially designed, high-strength, monodispersed silica particles, combined with well-established column packing processes. BioCore SEC-150 columns are suited for aggregate determination of peptides, small proteins and related substances.
Catalog No. NC3585536 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. B21303001504615S
BioCore SEC is a family of high performance, size exclusion chromatography columns designed for separating a broad range of biomolecules based on size. This column technology involves the creation of a neutral hydrophilic layer on the surface of specially designed, high-strength, monodispersed silica particles, combined with well-established column packing processes. BioCore SEC-150 columns are suited for aggregate determination of peptides, small proteins and related substances.