Learn More
NANOMICRO TECHNOLOGIES INC NanoChrom Technologies, BioCore WA x LC Analytical Column, 10um, 4.6 x 50 mm, PEEK
Catalog No. NC3587952
Encompass_Preferred
BioCore WAX is a family of high-performance, weak anion-exchange columns designed for separating charged variants in low pI proteins, such as fusion proteins. Its column technology involves the creation of a hydrophilic tertiary ammonium functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.
Catalog No. NC3587952 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. B51110000004605P
BioCore WAX is a family of high-performance, weak anion-exchange columns designed for separating charged variants in low pI proteins, such as fusion proteins. Its column technology involves the creation of a hydrophilic tertiary ammonium functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.