Learn More
NANOMICRO TECHNOLOGIES INC NanoChrom Technologies, BioCore WC x LC Preparative Column, 10um, 10 x 250 mm, Stainless Steel
Catalog No. NC3585551
Encompass_Preferred
BioCore WCX is a family of high-performance, weak cation-exchange columns designed for separating charged variants in proteins, including monoclonal antibodies (mAbs). Its column technology involves the creation of a hydrophilic carboxylic functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.
Catalog No. NC3585551 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. B31110000010025S
Add to Cart
Add to Cart
Request a Quote
BioCore WCX is a family of high-performance, weak cation-exchange columns designed for separating charged variants in proteins, including monoclonal antibodies (mAbs). Its column technology involves the creation of a hydrophilic carboxylic functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.