Learn More
Electron Microscopy Sciences BioCulture™ BioCulture Calibration Head
SDP
Catalog No. 5033876
Encompass
The BioCulture Calibration Head is A NIST traceable venture calibrator with 1% accuracy that allow users to calibrate the flow of the BioCulture pump themselves without having to send the pump out for service of calibration. It allows flow calibration at flow rates between 30-120 LPM. It is designed for use with impellar fan typle pumps like the BioCulture, which require calibration devices with very little backpressure resistance.
Catalog No. 50-338-76 Supplier Electron Microscopy Sciences Supplier No. 6522450
The BioCulture Calibration Head is A NIST traceable venture calibrator with 1% accuracy that allow users to calibrate the flow of the BioCulture pump themselves without having to send the pump out for service of calibration. It allows flow calibration at flow rates between 30-120 LPM. It is designed for use with impellar fan typle pumps like the BioCulture, which require calibration devices with very little backpressure resistance.