Learn More
Electron Microscopy Sciences BioCulture™ Microbial Air Sampler BioCulture Pump & Calibration Kit
SDP
Catalog No. 5033875
Encompass
This BioCulture pump features a selectable, continuous, constant sample flowrate and is simple and quiet to operate for a variety of applications. No tools are needed. Ideal for applications in IAQ surveys, mold sampling, building diagnostics, clean room sampling, hospital operating rooms, pharmaceutical compounding labs, USP 797 active air sampling compliance, and any location that needs to verify and quantify microorganism presence.
The flow rate is 30-120 LPM with an accuracy of +/-5%. Uses standard 90 mm petri dishes. Sampling head design includes 380 1 mm diameter holes and may be sterilized by autoclave or gas torch. NiCAD battery design that runs for 6.8 hours depending on flow rate, battery maintenance and back-pressure and recharge in 12 to 16 hours. The tripod mounting threads on bottom of unit allow for 90 degree sampling. Built-in timers of 1, 2, 5, 10 minutes. Includes pump, calibration head, standard 120V charger, pencil torch, carrying case & manual.
Catalog No. 50-338-75 Supplier Electron Microscopy Sciences Supplier No. 6522420
This BioCulture pump features a selectable, continuous, constant sample flowrate and is simple and quiet to operate for a variety of applications. No tools are needed. Ideal for applications in IAQ surveys, mold sampling, building diagnostics, clean room sampling, hospital operating rooms, pharmaceutical compounding labs, USP 797 active air sampling compliance, and any location that needs to verify and quantify microorganism presence.
The flow rate is 30-120 LPM with an accuracy of +/-5%. Uses standard 90 mm petri dishes. Sampling head design includes 380 1 mm diameter holes and may be sterilized by autoclave or gas torch. NiCAD battery design that runs for 6.8 hours depending on flow rate, battery maintenance and back-pressure and recharge in 12 to 16 hours. The tripod mounting threads on bottom of unit allow for 90 degree sampling. Built-in timers of 1, 2, 5, 10 minutes. Includes pump, calibration head, standard 120V charger, pencil torch, carrying case & manual.