Learn More
Abcam Biocytin hydrazide hydrochloride, 10MG
Catalog No. 50996074
Encompass_Preferred
MW 386.5 Da. Water-soluble, long-chained biotin hydrazide for glycoconjugate detection. Selective label for sialic acid, galactose and other sugars in glycoconjugates using avidin-biotin technology. Can biotin label proteins and peptides. Linking arm enables easier binding to avidin / sterptavidin in ELISA. Cellular tracer. Can be introduced into cells via micro-injection. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-074 Supplier Abcam Supplier No. AB27521410MG
MW 386.5 Da. Water-soluble, long-chained biotin hydrazide for glycoconjugate detection. Selective label for sialic acid, galactose and other sugars in glycoconjugates using avidin-biotin technology. Can biotin label proteins and peptides. Linking arm enables easier binding to avidin / sterptavidin in ELISA. Cellular tracer. Can be introduced into cells via micro-injection. .
The product is subject to the following: Abcam Restricted Use Statement