Learn More
Medchemexpress LLC HY-101884 10mg Medchemexpress, Biocytin CAS:576-19-2 Purity:>98%
SDP
Catalog No. 501967906
Encompass_Preferred
Medchemexpress, HY-101884 10mg Biocytin CAS:576-19-2 Biocytin is a conjugate of D-biotin and L-lysine, where the carboxylate of D-biotin is coupled with the ϵ-amine of L-lysine via a secondary amide bond. Biocytin is a classical neuroanatomical tracer commonly used to map brain connectivity. Biocytin is used as a versatile marker in anterograde, retrograde and intracellular neuroanatomical investigations and in biotinidase assays. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7906 Supplier Medchemexpress LLC Supplier No. HY10188410MG
Medchemexpress, HY-101884 10mg Biocytin CAS:576-19-2 Biocytin is a conjugate of D-biotin and L-lysine, where the carboxylate of D-biotin is coupled with the ϵ-amine of L-lysine via a secondary amide bond. Biocytin is a classical neuroanatomical tracer commonly used to map brain connectivity. Biocytin is used as a versatile marker in anterograde, retrograde and intracellular neuroanatomical investigations and in biotinidase assays. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.