Learn More
CPC Scientific Biotin-Lys-Tyr-Gly-Gly-Phe-Met-Thr-Ser-Glu-Lys-Ser-Gln-Thr-Pro-Leu-Val-Thr-Leu-Phe-Lys-Asn-Ala-Ile-Ile-Lys-Asn-Ala-Tyr-Lys-Lys-Gly-Glu-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific ENDP007A
Encompass
ONE-LETTER SEQUENCE: Biotin-KYGGFMTSEKSQTPLVTLFKNAIIKNAYKKGEMOLECULAR FORMULA: C174H277N43O49S2MOLECULAR WEIGHT:3818.44STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [135482-75-6], RESEARCH AREA: Pain
Catalog No. 50-210-9231