Learn More
Medchemexpress LLC Biocytin hydrazide | 102743-85-1 | 99.2% | 386.51 | 5 MG
SDP
Catalog No. 5000360224
Encompass_Preferred
Biocytin hydrazide is a selective label for sialic acids, galactose, and other sugars in glycoconjugates, utilizing avidin-biotin technology.
  • Selective label for sialic acids, galactose, and other sugars in glycoconjugates
  • Utilized with avidin-biotin technology
  • Intended for research use only
Catalog No. 50-003-60224 Supplier Medchemexpress LLC Supplier No. HY1471685MG
Biocytin hydrazide is a selective label for sialic acids, galactose, and other sugars in glycoconjugates, utilizing avidin-biotin technology.
  • Selective label for sialic acids, galactose, and other sugars in glycoconjugates
  • Utilized with avidin-biotin technology
  • Intended for research use only