Learn More
Biotium Biocytin Hydrazide
Catalog No. 501964814
Encompass_Preferred
Biocytin hydrazide is an aldehyde-fixable cellular tracer that can be introduced into cells by microinjection. Biocytin hydrazide can also be used to label glycoproteins or other carbohydrate molecules. Properties: White solid soluble in DMSO; Store desiccated at 4°C; C16H30N6O3S; MW: 386.51; [102743-85-1]
Catalog No. 50-196-4814 Supplier Biotium Supplier No. 90060
Add to Cart
Edge
Add to Cart
Biocytin hydrazide is an aldehyde-fixable cellular tracer that can be introduced into cells by microinjection. Biocytin hydrazide can also be used to label glycoproteins or other carbohydrate molecules. Properties: White solid soluble in DMSO; Store desiccated at 4°C; C16H30N6O3S; MW: 386.51; [102743-85-1]