Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Biocytin Hydrazide

Catalog No. B1603
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1603 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1603 Supplier Invitrogen™ Supplier No. B1603
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

biocytin hydrazide

Biocytin hydrazide an be used as microinjectable anterograde and transneuronal tracers that is fixable with aldehyde-based fixatives. Additionally, the primary aliphatic amine of this molecule can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3) to form new biotinylated probes. Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247).
TRUSTED_SUSTAINABILITY

Specifications

Product Type Biocytin Hydrazide
Quantity 25 mg
Content And Storage Store at room temperature.
Reagent Type Biocytins
Shipping Condition Room Temperature
Label Type Biotin & Other Haptens
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.