Learn More
Electron Microscopy Sciences Bio-Pure™ Biodegradable Sterile Reservoir, 50 ml volume
SDP
Catalog No. 5034538
Encompass_Preferred
A special additive helps the plastic bio-degrade by allowing bacteria and other organisms to breakdown the complex molecular chain. This speeds up the process to reduce the resin to its natural components within five years as opposed to 1,000 years or more. 100% Eco-Friendly - Each case of Bio-Pure Reservoirs are packed in bio-degradable bags and recycled cardboard cases.
Volume: 50 ml
Catalog No. 50-345-38 Supplier Electron Microscopy Sciences Supplier No. 7049748
A special additive helps the plastic bio-degrade by allowing bacteria and other organisms to breakdown the complex molecular chain. This speeds up the process to reduce the resin to its natural components within five years as opposed to 1,000 years or more. 100% Eco-Friendly - Each case of Bio-Pure Reservoirs are packed in bio-degradable bags and recycled cardboard cases.
Volume: 50 ml