Learn More
Diversified Biotech Inc Biodegradeable 6" Sampling spoon, 10cc level capacity 25/pack (Non Sterile)
Catalog No. NC3360883
Encompass
Planet-Safe Sampling spoons are 100% plant-based, made from PLA derived from sustainably grown, non-GMO sugarcane. They are eco-friendly with disposal options including commercial composting, incineration, or landfill, offering carbon sequestration benefits. Pack of 25.
Catalog No. NC3360883 Supplier Diversified Biotech Inc Supplier No. BPSP2000
Planet-Safe Sampling spoons are 100% plant-based, made from PLA derived from sustainably grown, non-GMO sugarcane. They are eco-friendly with disposal options including commercial composting, incineration, or landfill, offering carbon sequestration benefits. Pack of 25.