Learn More
Perkin Elmer US LLC Biodiesel Application Pack
Catalog No. 502117237
Encompass_Preferred
The Biodiesel Application Pack contains: Two-port Leur Lock %T cell with BaF2 windows Package of assorted Teflon spacers for the %T cell Two 2 mL Leur Lock syringes Liquid cell Leur connected waste tubing Fuels Touch Apps Software Kit The Touch Apps Software Kit includes the software and SOPs to support the analysis of biofuels and provide a simplified workflow-driven operating environment; for faster, more reproducible analyses.
Catalog No. 50-211-7237 Supplier Perkin Elmer US LLC Supplier No. L1608010
Add to Cart
Add to Cart
Request a Quote
The Biodiesel Application Pack contains: Two-port Leur Lock %T cell with BaF2 windows Package of assorted Teflon spacers for the %T cell Two 2 mL Leur Lock syringes Liquid cell Leur connected waste tubing Fuels Touch Apps Software Kit The Touch Apps Software Kit includes the software and SOPs to support the analysis of biofuels and provide a simplified workflow-driven operating environment; for faster, more reproducible analyses.