Learn More
Sigma Aldrich Fine Chemicals Biosciences Biodiesel ERMR certified r
Catalog No. 502900104
Encompass
Certified for ester content linolenic acid methyl ester content monoglyceride content diglyceride content triglyceride content total glycerol content water content density (at 15 C) viscosity (at 40 C) oxidation stability (at 110 C) acid value iodine value and flash point.
Catalog No. 50-290-0104 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. ERMEF00127ML
Certified for ester content linolenic acid methyl ester content monoglyceride content diglyceride content triglyceride content total glycerol content water content density (at 15 C) viscosity (at 40 C) oxidation stability (at 110 C) acid value iodine value and flash point.