Learn More
Seracare Life Sciences Inc BIODYNE B NYLON MEMBRANE
Supplier:  Seracare Life Sciences Inc 59600025
Encompass_Preferred
Biodyne B Membrane, manufactured by Pall, is positively charged nylon membrane with a pore size of 0.45 um. Biodyne B is specifically recommended for use with KPL Detector Nucleic Acid Detection Systems. Biodyne B membrane may be used to bind nucleic acids or protiens in such protocols as Southern, Northern, Western, or dot blotting.
Catalog No. 50-675-45
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.