Learn More
Medchemexpress LLC BioE-1115 | 1268863-35-9 | >98.2% | 1 ML
SDP
Catalog No. 5000339690
Encompass_Preferred
BioE-1115 is a highly selective and potent PAS kinase (PASK) inhibitor with an IC50 of ~4 nM. It also acts as a potent casein kinase 2α inhibitor with an IC50 of ~10 μM. This compound is intended for research use only.
  • Highly selective and potent PAS kinase inhibitor
  • Potent casein kinase 2α inhibitor
  • Reduces SREBP activity without affecting cell morphology or growth rate
  • Decreases insulin resistance, hepatic and serum TAG, and serum glucose in animal models
  • Suppresses expression of SREBP-1c target genes
Catalog No. 50-003-39690 Supplier Medchemexpress LLC Supplier No. HY12957110MM/1ML
BioE-1115 is a highly selective and potent PAS kinase (PASK) inhibitor with an IC50 of ~4 nM. It also acts as a potent casein kinase 2α inhibitor with an IC50 of ~10 μM. This compound is intended for research use only.
  • Highly selective and potent PAS kinase inhibitor
  • Potent casein kinase 2α inhibitor
  • Reduces SREBP activity without affecting cell morphology or growth rate
  • Decreases insulin resistance, hepatic and serum TAG, and serum glucose in animal models
  • Suppresses expression of SREBP-1c target genes