Learn More
Medchemexpress LLC BioE-1115 | 1268863-35-9 | 98.2% | 50 MG
SDP
Catalog No. 5000339445
Encompass_Preferred
BioE-1115 is a highly selective and potent PAS kinase (PASK) inhibitor with an IC50 of approximately 4 nM. It also acts as a potent casein kinase 2α inhibitor with an IC50 of about 10 μM.
  • Highly selective and potent PAS kinase (PASK) inhibitor
  • Potent casein kinase 2α inhibitor
  • Dose-dependent loss of PASK phosphorylation in HEK293 cells
  • Reduces SREBP activity in HepG2 cells
  • Suppresses Gpat1, Fasn, and other SREBP-1c target genes in rats
  • Decreases hepatic expression of lipogenic SREBP-1c target genes
  • Reduces serum triglycerides and partially reverses insulin resistance
  • Decreases HOMA-IR and significantly decreases serum glucose
Catalog No. 50-003-39445 Supplier Medchemexpress LLC Supplier No. HY12957150MG
BioE-1115 is a highly selective and potent PAS kinase (PASK) inhibitor with an IC50 of approximately 4 nM. It also acts as a potent casein kinase 2α inhibitor with an IC50 of about 10 μM.
  • Highly selective and potent PAS kinase (PASK) inhibitor
  • Potent casein kinase 2α inhibitor
  • Dose-dependent loss of PASK phosphorylation in HEK293 cells
  • Reduces SREBP activity in HepG2 cells
  • Suppresses Gpat1, Fasn, and other SREBP-1c target genes in rats
  • Decreases hepatic expression of lipogenic SREBP-1c target genes
  • Reduces serum triglycerides and partially reverses insulin resistance
  • Decreases HOMA-IR and significantly decreases serum glucose