Learn More
TARGETMOL CHEMICALS INC BIOE-1115 5MG
SDP
Catalog No. 502581709
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BioE-1115 is a selective and potent inhibitor of serine-threonine protein kinase (PASK IC50 = 4 nM). BioE-1115 is a potent inhibitor of CK2(alpha) (IC50 = 10 uM). purity: 97%
Catalog No. 50-258-1709 Supplier TARGETMOL CHEMICALS INC Supplier No. T105475MG
Also available in 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BioE-1115 is a selective and potent inhibitor of serine-threonine protein kinase (PASK IC50 = 4 nM). BioE-1115 is a potent inhibitor of CK2(alpha) (IC50 = 10 uM). purity: 97%