Learn More
Medchemexpress LLC HY-129571 5mg Medchemexpress, BioE-1115 CAS:1268863-35-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1295715MG
Encompass_Preferred
Medchemexpress, HY-129571 5mg BioE-1115 CAS:1268863-35-9 BioE-1115 is a highly selective and potent PAS kinase (PASK) inhibitor with an IC50 of ~4 nM. BioE-1115 is also a potent casein kinase 2α inhibitor with an IC50 of ~10 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0161