Learn More
Cambridge Isotope Laboratories BIOEXPRESS CELL GROWTH MEDIA 10X CONCENTRATE (U-13C,98%; U-15N, 98%; U-D 98%), 10 ML SAMP
Catalog No. 501629895
Encompass
BIOEXPRESS CELL GROWTH MEDIA 10X CONCENTRATE (U-13C,98%; U-15N, 98%; U-D 98%), 10 ML SAMP
Catalog No. 50-162-9895 Supplier Cambridge Isotope Laboratories Supplier No. CGM1000CDNS
BIOEXPRESS CELL GROWTH MEDIA 10X CONCENTRATE (U-13C,98%; U-15N, 98%; U-D 98%), 10 ML SAMP