Learn More
Cambridge Isotope Laboratories BIOEXPRESS CELL GROWTH MEDIA (U-D, 98%; U-15N, 98%) (10X CONCENTRATE), 10 ML SAMP
Catalog No. 501629904
Encompass
BIOEXPRESS CELL GROWTH MEDIA (U-D, 98%; U-15N, 98%) (10X CONCENTRATE), 10 ML SAMP
Catalog No. 50-162-9904 Supplier Cambridge Isotope Laboratories Supplier No. CGM1000DNS
BIOEXPRESS CELL GROWTH MEDIA (U-D, 98%; U-15N, 98%) (10X CONCENTRATE), 10 ML SAMP