Learn More
Chemglass Life Sciences Biofactory 1 Chamber, Wide Mouth, Total Culture Area: 647cm2, TC, Sterile, NEST, 8 Per Case
SDP
Catalog No. 501941124
Encompass_Preferred
NEST BioFactory™ system is a cell culture device that utilizes the maximum culture area in a limited space. NEST BioFactory™ system can be used for industrial scale production such as vaccine, monoclonal antibodies or biopharmaceuticals production, and are suitable for adherent cell culture. Each biofactory has the vent filter caps installed on the two ports but they are also supplied with a pkg of 2 solid caps per biofactory. Features: Wide mouth Double neck w/ Filter Vent Caps, Double-layer sterile bag, Clear lot number for batch traceability, Manufactured in dedicated 10,000-class clean room following GMP standards, Ultrasonic welding process without any chemical addition, Surface TC treatment ensures efficient cell attachment, In-house E-beam sterilization meets ISO13485 and ISO 11137:2006 standards and achieves sterility level SAL = 10-6, Compatible with Corning CellSTACK Culture Chamber accessories.
Catalog No. 50-194-1124 Supplier Chemglass Life Sciences Supplier No. CGN7710001
NEST BioFactory™ system is a cell culture device that utilizes the maximum culture area in a limited space. NEST BioFactory™ system can be used for industrial scale production such as vaccine, monoclonal antibodies or biopharmaceuticals production, and are suitable for adherent cell culture. Each biofactory has the vent filter caps installed on the two ports but they are also supplied with a pkg of 2 solid caps per biofactory. Features: Wide mouth Double neck w/ Filter Vent Caps, Double-layer sterile bag, Clear lot number for batch traceability, Manufactured in dedicated 10,000-class clean room following GMP standards, Ultrasonic welding process without any chemical addition, Surface TC treatment ensures efficient cell attachment, In-house E-beam sterilization meets ISO13485 and ISO 11137:2006 standards and achieves sterility level SAL = 10-6, Compatible with Corning CellSTACK Culture Chamber accessories.