Learn More
Alkali Scientific BioFactory Systems Chambers Ce
SDP
Catalog No. 503028480
Encompass_Preferred
BioFactory Systems are compact multi-layer single-use cell culture systems designed for easy scale-up cell culture applications such as production of vaccines monoclonal antibodies or pharmaceuticals Theyre versatile and easy-to-use system for medium to large scale research or commercial production with reduced contamination risk Narrow mouth caps available for tubing solutions Vented Overcap and Solid Overact can be used directly on the adaptor caps to block dust and bacteria Vented Overcap is equipped with a 0 22 m hydrophobic breathable membrane to achieve aseptic ventilation during liquid transfer Available in five sizes 1-Chamber with 647 cm2 cell growth area 2-Chamber with 1279 cm2 cell growth area 5-Chamber with 3175 cm2 cell growth area 10-Chamber with 6320 cm2 cell growth area 40-Chamber with 25280cm2 cell growth area Made of high clarity 100% virgin polystyrene Sterilized by E-beam SAL10-6 Non-Pyrogenic DNase/Rnase free Individually packaged in double-layer sterile
Catalog No. 50-302-8480 Supplier Alkali Scientific Supplier No. TV84
BioFactory Systems are compact multi-layer single-use cell culture systems designed for easy scale-up cell culture applications such as production of vaccines monoclonal antibodies or pharmaceuticals Theyre versatile and easy-to-use system for medium to large scale research or commercial production with reduced contamination risk Narrow mouth caps available for tubing solutions Vented Overcap and Solid Overact can be used directly on the adaptor caps to block dust and bacteria Vented Overcap is equipped with a 0 22 m hydrophobic breathable membrane to achieve aseptic ventilation during liquid transfer Available in five sizes 1-Chamber with 647 cm2 cell growth area 2-Chamber with 1279 cm2 cell growth area 5-Chamber with 3175 cm2 cell growth area 10-Chamber with 6320 cm2 cell growth area 40-Chamber with 25280cm2 cell growth area Made of high clarity 100% virgin polystyrene Sterilized by E-beam SAL10-6 Non-Pyrogenic DNase/Rnase free Individually packaged in double-layer sterile