Learn More
Alkali Scientific BioFactory Systems Chambers Ce
SDP
Supplier:  Alkali Scientific TV91
Encompass_Preferred
BioFactorySystems are compact multi-layer single-use cell culture systems designed for easy scale-up cell culture applications such as production of vaccines monoclonal antibodies or pharmaceuticals Theyre versatile and easy-to-use systems for medium to large scale research or commercial production with reduced contamination risk Narrow mouth caps available for tubing solutions Available in five sizes 1-Chamber with 632cm2 cell growth area 2-Chamber with 1264cm2 cell growth area 5-Chamber with 3160cm2 cell growth area 10-Chamber with 6335 cm2 cell growth area 40-Chamber with 25295 cm2 cell growth area Made of high clarity 100% virgin polystyrene Sterilized by E-beam SAL10-6 Non-Pyrogenic DNase/Rnase free Individually packaged in double-layer sterile bags TC treated good for cell attachment and growth Clear lot number for batch traceability Ultrasonic welding without extrinsic ingredients Excellent welding line design can avoid producing particles during welding and make the b
Catalog No. 50-302-8482