Learn More
Bioworld BioFectin™, 0.5 mL
Catalog No. 50255444
Encompass_Preferred
BioFectinTMis a newly developed second generation of lipid-based Transfection Reagent that is optimized for transfection in normal serum-containing medium reagents.It is a one-step solution for all transfection needs.It is highly effective with DNA and siRNA transfection.Features:- For High efficiency DNA and siRNA transfection- Low toxicity- very broad nucleic acid to reagent ratios, no need to optimize conditions- Works in serum-containing medium Appearance: Liquid
Catalog No. 50-255-444 Supplier Bioworld Supplier No. 402200121
BioFectinTMis a newly developed second generation of lipid-based Transfection Reagent that is optimized for transfection in normal serum-containing medium reagents.It is a one-step solution for all transfection needs.It is highly effective with DNA and siRNA transfection.Features:- For High efficiency DNA and siRNA transfection- Low toxicity- very broad nucleic acid to reagent ratios, no need to optimize conditions- Works in serum-containing medium Appearance: Liquid