Learn More
Braintree Scientific BioFree Pipette Tips, 1000ul max, bulk pack of 5,000
SDP
Catalog No. 501955432
Encompass
Labnet BioFree pipette pips are designed to fit a wide variety of single and multi-channel pipettes. Flexible walls and a series of internal sealing rings ensure a secure fit and minimal force to eject. Tips are manufactured from high quality, FDA compliant poly propylene in a clean room environment. Diamond polished, stainless steel molds produce a tip with exceptionally smooth surfaces and extreme accuracy. During manufacturing process, each lot of tips is tested for accuracy and precision. Tips are also carefully inspected for uniformity and manufacturing irregularities. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5432 Supplier Braintree Scientific Supplier No. P10001000
Labnet BioFree pipette pips are designed to fit a wide variety of single and multi-channel pipettes. Flexible walls and a series of internal sealing rings ensure a secure fit and minimal force to eject. Tips are manufactured from high quality, FDA compliant poly propylene in a clean room environment. Diamond polished, stainless steel molds produce a tip with exceptionally smooth surfaces and extreme accuracy. During manufacturing process, each lot of tips is tested for accuracy and precision. Tips are also carefully inspected for uniformity and manufacturing irregularities. **Sold for research purposes only and not for human use**