Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Biofuel from Alcohol Fermentation Experiment Kit
SDPSCI_ED

Catalog No. s29015 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S29015 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S29015 Supplier Edvotek™ Supplier No. 304
Only null left
Add to Cart
Add to Cart

Requires

Air pump, heater, pH probe, temperature probe, ethanol sensor, fermentor vessel, autoclave, ice, shaker, ethanol, DH20, centrifuge, colorimeter.

Designed to introduce basic concepts of fermentation and bioprocessing through the production of Ethanol in a small-scale fermentor. Edvotek™ Biofuel from Alcohol Fermentation Experiment Kit uses Saccharomyces cerevisiae to break down sugar into ethanol.

  • Experiment conditions controlled with mechanical devices are aeration and temperature
  • pH, bacterial growth, and ethanol production are monitored using specific probes over a period of 5 hours
  • For 10 Lab Groups
  • Complete in 5-6 hours
  • Level: Advanced

Specifications

Grade 9 to Undergraduate
Includes Instructions, Yeast MicroBead, yeast growth medium, syringe, 15mL centrifuge tubes, transfer pipettes, loops.
Time Range Complete in 5-6 hours
Content And Storage Refrigerator (some components)
Class Size 10 Lab Groups
Product Type Biofuel from Alcohol Fermentation Experiment Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.