Learn More
MSC Rochester Midland Biogenic SE 374, Liquid, D-Limonene, 5 gal
Catalog No. 502870460
Encompass
Biogenic SE 374 is a neutral, concentrated blend of d-Limonene, biodegradable hydrocarbon solvents, and synthetic detergent. This liquid adhesive remover is designed for rapid penetration of oil, grease, tar, and asphalt. It forms a milky, partially stable emulsion with water.
  • Highly concentrated
  • Safe on all metals, glass, and concrete
  • Biodegradable
  • Contains no petroleum, aromatic, or chlorinated solvents
  • Non-phosphated
  • No alkaline builders - neutral pH
  • Effectively removes asphalt, oil, grease, and tar
  • Applicable via coarse spray, wipe, or cold dip tank
  • Light yellow liquid with citrus fragrance
  • Thin viscosity
Catalog No. 50-287-0460 Supplier MSC Supplier No. 81216285
Biogenic SE 374 is a neutral, concentrated blend of d-Limonene, biodegradable hydrocarbon solvents, and synthetic detergent. This liquid adhesive remover is designed for rapid penetration of oil, grease, tar, and asphalt. It forms a milky, partially stable emulsion with water.
  • Highly concentrated
  • Safe on all metals, glass, and concrete
  • Biodegradable
  • Contains no petroleum, aromatic, or chlorinated solvents
  • Non-phosphated
  • No alkaline builders - neutral pH
  • Effectively removes asphalt, oil, grease, and tar
  • Applicable via coarse spray, wipe, or cold dip tank
  • Light yellow liquid with citrus fragrance
  • Thin viscosity