Learn More
Research Products International Corp Red Color Polyethylene Bag Printed with BioHazard Symbol, 2 mil Thick, 12 x 18 Inch Bags, 100 per Package
SDP
Catalog No. 50998762
Encompass
Bright red, bags feature one long flap for easy opening. Printed with the biohazard symbol and sterilization indicator patch, they are made of High Molecular Weight, High-Density (HMHD) polyethylene. Ensure safe handling and disposal of biohazardous waste. Fits a variety of containers and holders.
Catalog No. 50-998-762 Supplier Research Products International Corp Supplier No. 246105
Bright red, bags feature one long flap for easy opening. Printed with the biohazard symbol and sterilization indicator patch, they are made of High Molecular Weight, High-Density (HMHD) polyethylene. Ensure safe handling and disposal of biohazardous waste. Fits a variety of containers and holders.