Learn More
Research Products International Corp Red Color Polyethylene Bag Printed with Biohazard Symbol, 2 mil Thick, 24 x 24 Inch Bags, 50 per Package
SDP
Catalog No. 50998763
Encompass
Bright red, bags feature one long flap for easy opening. Printed with the biohazard symbol and sterilization indicator patch, they are made of High Molecular Weight, High-Density (HMHD) polyethylene. Ensure safe handling and disposal of biohazardous waste. Fits a variety of containers and holders. Bags hold up to 8 gallons.
Catalog No. 50-998-763 Supplier Research Products International Corp Supplier No. 246106
Bright red, bags feature one long flap for easy opening. Printed with the biohazard symbol and sterilization indicator patch, they are made of High Molecular Weight, High-Density (HMHD) polyethylene. Ensure safe handling and disposal of biohazardous waste. Fits a variety of containers and holders. Bags hold up to 8 gallons.