Learn More
Uline Biohazard Bags 200/Case
Catalog No. NC1962848
Encompass
Red 1.2 mil thick polyethylene biohazard waste bag with biohazard symbol print, 30 IN x 36 IN (76 CM x 91 CM), 20-30 GL (75.7-113.6 LT) capacity for containing infectious or biologically hazardous waste for proper disposal, sold in cases of 250 EA.
Catalog No. NC1962848 Supplier Uline Supplier No. S20849
Red 1.2 mil thick polyethylene biohazard waste bag with biohazard symbol print, 30 IN x 36 IN (76 CM x 91 CM), 20-30 GL (75.7-113.6 LT) capacity for containing infectious or biologically hazardous waste for proper disposal, sold in cases of 250 EA.