Learn More
Research Products International Corp Autoclavable BioHazard Bags, Printed, 12 x 24 Inches, 100 per Package
SDP
Catalog No. 50998755
Encompass
Great economical solution for safe disposal of used pipettes, Petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters and other biohazard wastes.
•    Transparent polypropylene; 0.03mm (1.5 mil) thick
•    High temperature autoclavable at a maximum of 135˚C (275˚F)
•    Plain, non-printed bags can also be used as conventional trash can liners
•    Sterilization indicator patch darkens when exposed to steam sterilization for 20 minutes
•    Bags with biohazard warning label contain standard biohazard warning symbol and precautionary procedures in four languages: English, French, German, and Spanish
This item is not returnable. View return policy
Catalog No. 50-998-755 Supplier Research Products International Corp Supplier No. F131600009
Great economical solution for safe disposal of used pipettes, Petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters and other biohazard wastes.
•    Transparent polypropylene; 0.03mm (1.5 mil) thick
•    High temperature autoclavable at a maximum of 135˚C (275˚F)
•    Plain, non-printed bags can also be used as conventional trash can liners
•    Sterilization indicator patch darkens when exposed to steam sterilization for 20 minutes
•    Bags with biohazard warning label contain standard biohazard warning symbol and precautionary procedures in four languages: English, French, German, and Spanish