Learn More
MTC Bio Bowtie™ Biohazard bags, autoclave bag with drawstring, PE, 33 x 41in. (84 x 104cm), with marking area and sterilization indicator, 100/pk
Catalog No. NC3345649
Encompass_Preferred
The exclusive drawstring loops on BowTie™ Biohazard Bags provide instant-cinch closure on your waste materials and a handle to easily remove bags from their bins. They also allow venting during sterilization and provide an easy, safe way to remove bags from a hot autoclave. BowTie™ Bag’s 2.25 mil thick HDPE is suitable for autoclaving in 121°C cycles. BowTie™ Bags are printed with a number of identifying markers such as large biohazard symbols and temperature limit warnings. They are also outfitted with a large marking area and a sterilization patch that changes color after autoclaving.BowTie™ Bags fit their respective biohazard bins perfectly. The hands-free lid offers quick disposal and a cleaner, touchless experience with just a simple step on the pedal. The smaller A8000B bin couples with 12 gallon bags while the larger A8002B XL bin is a perfect fit for new 30 gallon BowTie™ Bags.
Catalog No. NC3345649 Supplier MTC Bio Supplier No. A8003R
The exclusive drawstring loops on BowTie™ Biohazard Bags provide instant-cinch closure on your waste materials and a handle to easily remove bags from their bins. They also allow venting during sterilization and provide an easy, safe way to remove bags from a hot autoclave. BowTie™ Bag’s 2.25 mil thick HDPE is suitable for autoclaving in 121°C cycles. BowTie™ Bags are printed with a number of identifying markers such as large biohazard symbols and temperature limit warnings. They are also outfitted with a large marking area and a sterilization patch that changes color after autoclaving.BowTie™ Bags fit their respective biohazard bins perfectly. The hands-free lid offers quick disposal and a cleaner, touchless experience with just a simple step on the pedal. The smaller A8000B bin couples with 12 gallon bags while the larger A8002B XL bin is a perfect fit for new 30 gallon BowTie™ Bags.