Learn More
Grainger Biohazard Bags, 1 gal, Polyethylene, Red, Biohazard, PK 100
Supplier:  Grainger 3UAF6
Encompass_Preferred
Bags for biohazard waste are used in hospitals and other healthcare facilities to safely collect and dispose of hazardous medical waste such as linen, blood products, and materials contaminated with bodily fluids. These plastic bags are printed with words and symbols to identify the contents and encourage proper handling and disposal, protecting workers from unnecessary exposure and ensuring disposal procedures are followed. Use these bags in stands, receptacles, and bag racks to keep them open. Hazardous waste bags and transport bags are used to collect infectious medical waste, as well as waste that contains heavy metals, toxic materials, and chemicals that require special handling. Hazardous waste bags are used in hospitals, nursing homes, and industrial settings where hazardous waste is produced.
Catalog No. NC1821429