Learn More
Grainger FIRST AID ONLY Biohazard Bags, 20 gal Capacity
Catalog No. NC2150716
Encompass_Preferred
The First Aid Only Biohazard Bags are 20 gallon capacity bags designed for the disposal of biohazardous waste. They are made from 1.5 mil polyethylene and measure 32" x 40". The bags feature a pre-printed biohazard symbol and wording. They can be used for disposing of medical or biological waste. The bags meet OSHA standards for the handling of potentially infectious materials. They are sold in packs of 25 bags.
This item is not returnable. View return policy
Catalog No. NC2150716 Supplier Grainger Supplier No. 39P017
The First Aid Only Biohazard Bags are 20 gallon capacity bags designed for the disposal of biohazardous waste. They are made from 1.5 mil polyethylene and measure 32" x 40". The bags feature a pre-printed biohazard symbol and wording. They can be used for disposing of medical or biological waste. The bags meet OSHA standards for the handling of potentially infectious materials. They are sold in packs of 25 bags.