Learn More
MTC Bio BenchBin 3L™ Benchtop Biohazard Bin, 3 Liter Capacity, with Spring Loaded Lid, 6 x 5 x 8.5in, 1/ea
Catalog No. NC3345632
Encompass_Preferred
BenchBin™ is a personal waste container that any researcher can benefit from. Smaller autoclave bags to dispose of biological waste for subsequent processing or forget the autoclave bag and use the BenchBin as a waste container for non-hazardous waste. The red cover is easily removed from the inner basin for easy emptying of contents. Perfect for pipette, tips, microcentrifuge tubes, and gloves, the BenchBin features a push to open/close lid that keeps materials covered and your bench neat.
Catalog No. NC3345632 Supplier MTC Bio Supplier No. A8403B
BenchBin™ is a personal waste container that any researcher can benefit from. Smaller autoclave bags to dispose of biological waste for subsequent processing or forget the autoclave bag and use the BenchBin as a waste container for non-hazardous waste. The red cover is easily removed from the inner basin for easy emptying of contents. Perfect for pipette, tips, microcentrifuge tubes, and gloves, the BenchBin features a push to open/close lid that keeps materials covered and your bench neat.