Learn More
MTC Bio Biohazard Bin XL, large, 26 gallon capacity (100L), with foot pedal operated lid, 19 x 19 x 33in. / 48 x 48 84cm (W x L x H), 1/ea
Catalog No. NC3345655
Encompass
This full-size biohazard bin is perfect for all of the infectious or contaminated waste produced in laboratories on a daily basis. Clearly marked with the international biohazard symbol, there is no need for additional stickers or labels. The foot-pedal operated allows for hand free operation and reduces contamination risks. Notch hooks on the outside of the basket rim make installing bags simple and easy.
Catalog No. NC3345655 Supplier MTC Bio Supplier No. A8002B
This full-size biohazard bin is perfect for all of the infectious or contaminated waste produced in laboratories on a daily basis. Clearly marked with the international biohazard symbol, there is no need for additional stickers or labels. The foot-pedal operated allows for hand free operation and reduces contamination risks. Notch hooks on the outside of the basket rim make installing bags simple and easy.