Learn More
Alkali Scientific ASI™ Biohazard Bin XL, 23 Gallon
SDP
Catalog No. 503026502
Encompass_Preferred
This full-size biohazard bin is perfect for all of the infectious or contaminated waste produced in laboratories on a daily basis. Clearly marked with the international biohazard symbol, there is no need for additional stickers or labels. The foot-pedal operated allows for hand free operation and reduces contamination risks. Notch hooks on the outside of the basket rim make installing bags simple and easy.   • 23 gallon capacity• Touchless foot pedal operated lid• Printed biohazard symbol• Bag holding notches.
Catalog No. 50-302-6502 Supplier Alkali Scientific Supplier No. A3002B
This full-size biohazard bin is perfect for all of the infectious or contaminated waste produced in laboratories on a daily basis. Clearly marked with the international biohazard symbol, there is no need for additional stickers or labels. The foot-pedal operated allows for hand free operation and reduces contamination risks. Notch hooks on the outside of the basket rim make installing bags simple and easy.   • 23 gallon capacity• Touchless foot pedal operated lid• Printed biohazard symbol• Bag holding notches.