Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Biohazard Clamp On Shield
SDP

Catalog No. 22000216
Change view
Click to view available options
Width (English):
12 in.
Length (English):
8.5 in.
12 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English) Length (English)
22-000-216 12 in. 8.5 in.
22-000-217 12 in. 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-000-216 Supplier Poltex Supplier No. 8.5FLEXBIOCL
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Protect user from splash and biohazard exposure. Poltex Clamp On Biohazard Shield is made from polycarbonate.

  • Mounts to virtually any surface with a powerful clamp
  • Made from abrasion and break- resistant polycarbonate
  • Flexible neck for maximum adjustability
  • Available in two size: 8.5 x 12 in. and 12 x 12 in.

Specifications

Dimensions (L x W) 8.5 x 12 in.
Width (Metric) 30.5 cm
Width (English) 12 in.
Product Type Biohazard Clamp-on Shield
Length (English) 8.5 in.
Material Polycarbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.