Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Biohazard Disposal Bags
SDP

Catalog No. 22446145 Shop All Mopec Products
Change view
Click to view available options
Dimensions (L x W):
24 x 24 in.
33 x 39 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-446-145 24 x 24 in.
22-446-087 33 x 39 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-446-145 Supplier Mopec Supplier No. FS052
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Economical bags designed for safe infectious waste disposal and easy identification.

  • Designed for safe containment of infectious waste
  • Made of 0.002 in. polyethylene material
  • Clearly labeled in English and Spanish with the universal biohazard symbol

Specifications

Dimensions (L x W) 24 x 24 in.
Product Type Evidence Bag
Color Red with Black Print
Quantity 250/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.