Learn More
Sigma Aldrich Fine Chemicals Biosciences Biohazard disposal bags H
Catalog No. 502901183
Encompass
for personal or general lab use sizes to suit all needsA range of biohazard collection bags printed with biohazard warning symbol and precautions in English Spanish and frenchIncludes an indicator patch that darkens when autoclavedAutoclavable to 250f (121C)Disposable Epoxy coated steel wire holder available HS10044H
Catalog No. 50-290-1183 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. HS10320
for personal or general lab use sizes to suit all needsA range of biohazard collection bags printed with biohazard warning symbol and precautions in English Spanish and frenchIncludes an indicator patch that darkens when autoclavedAutoclavable to 250f (121C)Disposable Epoxy coated steel wire holder available HS10044H