Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Biohazard Disposal Bags with or without Warning Label

Catalog No. 13717287 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Dimensions (L x W):
24 x 12 in. (61 x 30 cm)
30 x 24 in. (76 x 61 cm)
36 x 24 in. (91 x 61 cm)
Color:
Clear
Translucent
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Color
13-717-287 24 x 12 in. (61 x 30 cm) Translucent
13-717-288 30 x 24 in. (76 x 61 cm) Translucent
22-032926 36 x 24 in. (91 x 61 cm) Translucent
13-717-289 24 x 12 in. (61 x 30 cm) Clear
13-717-290 30 x 24 in. (76 x 61 cm) Clear
13-717-291 36 x 24 in. (91 x 61 cm) Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 13-717-287 Supplier Bel-Art™ Supplier No. F131600005
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Economical, safe disposal of used pipettes, petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters, and other biohazard wastes

  • These transparent, polypropylene plastic biohazard bags are 0.03 mm (1.5 mil) thick
  • High temperature autoclavable at a maximum of 135°C (275°F)
  • May also be used as conventional trash can liners

Specifications

Capacity (Metric) 15 L
Certifications/Compliance ASTM D1709-98
Closure Type Zip Tie
Design Non-printed without Sterilization Patch
Capacity (English) 3 gal.
Color Translucent
Dimensions (L x W) 24 x 12 in. (61 x 30 cm)
Disposable Single-use
Length (Metric) 61 cm
Sterility Non-sterile
Width (Metric) 30 cm
Autoclavable Autoclavable
For Use With (Application) Disposal of used pipettes, petri dishes, culture plates, membrane filters and more
Width (English) 12 in.
Length (English) 24 in.
Thickness 1.5 mil (0.03 mm)
Material Polypropylene
Product Type Biohazard Bag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.