Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Biohazard Disposal Can
SCI_ED

Catalog No. s96727 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Capacity (English):
4 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
S96727 4 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S96727 Supplier Bel-Art™ Supplier No. F131970000/EMD
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ideal for biological/biohazard materials, broken glass, recyclables or other waste.

Autoclave bags (19 × 23 in. [48.3 × 58.4cm]) fit perfectly over a stainless-steel internal wire frame to keep bags open for safe and easy access.

  • Polypropylene construction
  • Puncture resistant
  • Bright red color for high visibility
  • For biohazard disposal
  • Autoclavable at 250°F (121°C)
  • Dimensions: 9 × 12 × 11.5 in. (22.5 × 30.5 × 29cm)

Specifications

Capacity (English) 4 gal.
Color Red
Dimensions (L x W x H) 9 x 12 x 11.5 in. (22.5 x 30.5 x 29 cm)
Height (Metric) 29 cm
Length (Metric) 22.5 cm
Width (Metric) 30.5 cm
Autoclavable Autoclavable
For Use With (Application) Biological or biohazard materials, broken glass, recyclables or other waste and sized for benchtop or floor placement
Width (English) 12 in.
Length (English) 9 in.
Height (English) 11.5 in.
Material Polypropylene
Product Type Biohazard Can
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.