Learn More
Research Products International Corp Paperboard Biohazard Sharp Object Disposal Pouch, 200 per Package
SDP
Catalog No. 50998777
Encompass
This hard to pierce pouch offers protection against the danger of being cut by contaminated sharp objects. Remains open in an oval shape by folding the bottom flaps. After filling, the top flaps close and lock in place for safe disposal. Printed with bold biohazard symbol.
Catalog No. 50-998-777 Supplier Research Products International Corp Supplier No. H13234
This hard to pierce pouch offers protection against the danger of being cut by contaminated sharp objects. Remains open in an oval shape by folding the bottom flaps. After filling, the top flaps close and lock in place for safe disposal. Printed with bold biohazard symbol.