Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitchell Plastics Biohazard Floor Waste Containers

Catalog No. 05723429
Change view
Click to view available options
Height (English):
29 in.
30.25 in.
Length (English):
14 in.
17.25 in.
Width (English):
14 in.
17.25 in.
For Use With (Equipment):
30.48 x 30.48 x 68.58 cm Box
91.44 x 114.3 cm Bag
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English) Length (English) Width (English) For Use With (Equipment)
05-723-429 29 in. 14 in. 14 in. 30.48 x 30.48 x 68.58 cm Box
05-723-426 30.25 in. 17.25 in. 17.25 in. 91.44 x 114.3 cm Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-723-429 Supplier Mitchell Plastics Supplier No. WB500

missing translation for 'validMainframeMsg'

Clear containers facilitate easy and safe segregation of hazardous materials for later disposal

  • Made of heavy-duty acrylic to help protect personnel from exposure to potential hazards
  • Clear design enables warning insignias and biohazard bag colors to be seen
  • Each unit has a hinged lid for easy placement of waste
  • Lids are also removable for easy positioning and removal of bag or box
  • Reusable

Floor models

  • Feature removable bottoms and can be used for radioactive material disposal
  • WC-100: holds bag 91.44 x 114.3cm (36 x 45 in.)
  • WB-500: holds box 30.48 x 30.48 x 68.58cm (12 x 12 x 27 in.)

Specifications

Color Clear
Dimensions (L x W x H) 14 x 14 x 29 in. (35.6 x 35.6 x 73.66 cm)
Height (Metric) 73.66 cm
Length (Metric) 35.6 cm
Model Floor
Width (Metric) 35.6 cm
For Use With (Application) For radioactive material
For Use With (Equipment) 30.48 x 30.48 x 68.58 cm Box
Width (English) 14 in.
Length (English) 14 in.
Thickness 12.7 mm
Height (English) 29 in.
Material Plastic
Product Type Waste Box Holder
Show More Show Less

Dispose contents according to all local, state and federal disposal regulations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.