Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDC Biohazard Hazard Label, Fluorescent Red, 500 Labels/Roll

Catalog No. 21235785
Change view
Click to view available options
Quantity:
500 Labels/Roll
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-235-785 500 Labels/Roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-235-785 Supplier PDC Supplier No. BH405
Add to Cart
Add to Cart

Enhance safety and visibility with PDC™ Biohazard Hazard Labels in bright fluorescent red for clear hazard identification.

  • Supplied on a 1 in. core; 500 labels per roll
  • Durable paper with permanent adhesive for secure application

Specifications

Adhesive Type Permanent
Coating None
Color Red
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Hazard Label
Sterility Non-sterile
Quantity 500 Labels/Roll
Dimensions (L x W) 3 x 2 in.
Length (English) 3 in.
Width (English) 2 in.
Width (Metric) 50.8 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.