Learn More
Description
- The Biohazard Keeper Floor Model offers a sturdy and economical solution for collecting non-sharp biohazardous lab waste. Designed for easy use and odor control
- Wide-mouth Reclose-A/Remove-A-Lid for easy disposal and odor reduction
- Durable corrugated construction with a 2 mil red plastic liner for leak resistance
- Ideal for petri dishes, tubes, plates, pipets, and specimen cups
- Clearly marked with orange biohazard symbols and bilingual safety warnings
Specifications
Specifications
| Capacity (Metric) | 63 L |
| Capacity (English) | 16.64 gal. |
| Color | White, Blue |
| Dimensions (L x W x H) | 30.5 x 30.5 x 68.6 cm (12 x 12 x 27 in.) |
| Disposable | Single-use |
| Model | Floor Model |
| Quantity | 6/Cs. |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| Material | Corrugated Cardboard |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.