Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Biohazard Labels
SDP

Catalog No. 22446068 Shop All Mopec Products
Change view
Click to view available options
Dimensions (L x W):
1.25 x 1 in.
8.5 x 11 in.
1 x 1 in.
1 x 4 in.
2 x 1.8 in.
1 x 1296 in.
Product Type:
Biohazard Continuous Tape
Biohazard Label
Biohazard Warning Tape
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Product Type
22-446-068 1 x 4 in. Biohazard Label
22-445-973 1 x 1 in. Biohazard Label
22-445-987 1.25 x 1 in. Biohazard Label
22-445-985 1 x 1296 in. Biohazard Continuous Tape
22-445-986 1 x 1296 in. Biohazard Warning Tape
22-446-002 2 x 1.8 in. Biohazard Label
22-446-056 8.5 x 11 in. Biohazard Label
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 22-446-068 Supplier Mopec Supplier No. FS338
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to identify evidence that may contain infectious body fluids and warn of health risks.

  • Features the international biohazard symbol for clear identification.
  • Available in multiple adhesive formats, including labels and roll tape.
  • Peel-away backing for labels
  • Fluorescent orange
  • Self-adhesive stock

Specifications

Adhesive Type Self-adhesive
Color Black on Red
Product Type Biohazard Label
Quantity 250/Roll
Dimensions (L x W) 1 x 4 in.
Length (English) 1 in.
Width (English) 4 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.